Learn More
Medchemexpress LLC Nsc23005 sodium | 1796596-46-7 | MFCD30534395 | 99.5% | 305.33 g/mol | C13H16NNaO4S | 1 ML
SDP
Catalog No. 5000212538
Encompass_Preferred
NSC23005 sodium is a small-molecule p18 inhibitor used in research to promote hematopoietic stem cell (HSC) expansion in murine and human systems. It is supplied as a 10 mM solution in DMSO or as a solid and is characterized by high purity and defined storage recommendations for laboratory use.
  • Promotes hematopoietic stem cell expansion in murine and human models.
  • Potent p18 inhibition with reported ED50 of 5.21 nM.
  • High purity (99.52%) suitable for research applications.
  • Available as a 10 mM solution in DMSO and in solid milligram quantities.
  • Store sealed at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-12538 Supplier Medchemexpress LLC Supplier No. HY10079110MM 1ML
NSC23005 sodium is a small-molecule p18 inhibitor used in research to promote hematopoietic stem cell (HSC) expansion in murine and human systems. It is supplied as a 10 mM solution in DMSO or as a solid and is characterized by high purity and defined storage recommendations for laboratory use.
  • Promotes hematopoietic stem cell expansion in murine and human models.
  • Potent p18 inhibition with reported ED50 of 5.21 nM.
  • High purity (99.52%) suitable for research applications.
  • Available as a 10 mM solution in DMSO and in solid milligram quantities.
  • Store sealed at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.