Learn More
Selleck Chemical LLC NSC232003-E0351-5MG
SDP
Catalog No. 502390070
Encompass_Preferred
NSC232003 is a highly potent and cell-permeable UHRF1 inhibitor that binds to the 5mC binding pocket of the SRA domain of UHRF1 NSC232003 modulates DNA methylation in a cellular context
Catalog No. 50-239-0070 Supplier Selleck Chemical LLC Supplier No. E03515MG
NSC232003 is a highly potent and cell-permeable UHRF1 inhibitor that binds to the 5mC binding pocket of the SRA domain of UHRF1 NSC232003 modulates DNA methylation in a cellular context