Learn More
Medchemexpress LLC NSC243928 (mesylate) | 59988-01-1 | 99.3% | 503.59 g/mol | C23H25N3O6S2 | 10MG

Supplier: Medchemexpress LLC HY12388510MG
NSC243928 mesylate is a small-molecule binder of lymphocyte antigen 6 (LY6) that inhibits cell growth and exhibits anticancer activity. The compound is provided as the mesylate salt, with a reported high purity and defined molecular formula and weight, and is intended for research use under controlled laboratory conditions.
- Small-molecule LY6 binder with reported anticancer activity.
- Inhibits cell growth in relevant assays.
- High reported purity (approximately 99.3%).
- Molecular formula C23H25N3O6S2 and molecular weight 503.59 g/mol.
- Soluble in DMSO at about 10 mg/mL (may require warming or sonication).
- Stable as a sealed powder when stored at 4°C and protected from moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.