Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NSC243928 mesylate | 59988-01-1 | 99.3% | 503.59 g/mol | C23H25N3O6S2 | 10MM 1ML
SDP

Catalog No. 5000235222
Encompass_Preferred

NSC243928 mesylate is a small-molecule LY6 (lymphocyte antigen 6) binder with reported antiproliferative and anticancer activity. It is available as a solid and as a ready-to-use 10 mM solution in DMSO for cell-based assays; published data report activity against several breast and ovarian cancer cell lines.

  • Binds LY6 family proteins and inhibits cell growth.
  • Reported IC50s: MDA-MB-231 5.683 μM; Hs-578-T 7.309 μM; BT-549 7.954 μM; OVCAR-8 4.167 μM.
  • Available as solid and 10 mM solution in DMSO (1 mL).
  • High purity: 99.33%.
  • Molecular formula: C23H25N3O6S2; molecular weight: 503.59 g/mol.
  • Soluble in DMSO to 10 mg/mL (≈19.86 mM) with warming.
  • Store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-35222 Supplier Medchemexpress LLC Supplier No. HY12388510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NSC243928 mesylate is a small-molecule LY6 (lymphocyte antigen 6) binder with reported antiproliferative and anticancer activity. It is available as a solid and as a ready-to-use 10 mM solution in DMSO for cell-based assays; published data report activity against several breast and ovarian cancer cell lines.

  • Binds LY6 family proteins and inhibits cell growth.
  • Reported IC50s: MDA-MB-231 5.683 μM; Hs-578-T 7.309 μM; BT-549 7.954 μM; OVCAR-8 4.167 μM.
  • Available as solid and 10 mM solution in DMSO (1 mL).
  • High purity: 99.33%.
  • Molecular formula: C23H25N3O6S2; molecular weight: 503.59 g/mol.
  • Soluble in DMSO to 10 mg/mL (≈19.86 mM) with warming.
  • Store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.