Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NSC243928 mesylate | 59988-01-1 | 99.3% | 503.59 g·mol⁻¹ | C23H25N3O6S2 | 5MG
SDP

Catalog No. 5000237981
Encompass_Preferred

NSC243928 mesylate is the mesylate salt of NSC243928, a small-molecule binder of human lymphocyte antigen 6 (LY6) that inhibits cell growth and exhibits anticancer activity in vitro. Reported activity includes induction of cell death and IC50 values in the low micromolar range against multiple cancer cell lines.

  • Solid, light yellow to yellow physical form.
  • Purity 99.33%.
  • Molecular weight 503.59 g·mol⁻¹ and formula C23H25N3O6S2.
  • Soluble in DMSO (10 mg/mL; may require sonication and warming).
  • Recommended storage: solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Provided in small research pack sizes and ready-to-use 10 mM DMSO solutions.

Catalog No. 50-002-37981 Supplier Medchemexpress LLC Supplier No. HY1238855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NSC243928 mesylate is the mesylate salt of NSC243928, a small-molecule binder of human lymphocyte antigen 6 (LY6) that inhibits cell growth and exhibits anticancer activity in vitro. Reported activity includes induction of cell death and IC50 values in the low micromolar range against multiple cancer cell lines.

  • Solid, light yellow to yellow physical form.
  • Purity 99.33%.
  • Molecular weight 503.59 g·mol⁻¹ and formula C23H25N3O6S2.
  • Soluble in DMSO (10 mg/mL; may require sonication and warming).
  • Recommended storage: solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Provided in small research pack sizes and ready-to-use 10 mM DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.