Learn More
Medchemexpress LLC HY-18931A 5mg Medchemexpress, NSC305787 (hydrochloride) CAS:53868-26-1 Purity:>98%
SDP
Catalog No. 502082091
Encompass_Preferred
Medchemexpress, HY-18931A 5mg NSC305787 (hydrochloride) CAS:53868-26-1 NSC305787 hydrochloride is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCΙ with an IC50 of 8.3 μM, has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2091 Supplier Medchemexpress LLC Supplier No. HY18931A5MG
Medchemexpress, HY-18931A 5mg NSC305787 (hydrochloride) CAS:53868-26-1 NSC305787 hydrochloride is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCΙ with an IC50 of 8.3 μM, has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.