Learn More
Medchemexpress LLC NSC319726 (ZMC1) | 71555-25-4 | 99.6% | 234.32 | 100 MG
SDP
Catalog No. 5000387865
Encompass_Preferred
NSC319726 (ZMC1) is a mutant p53R175 reactivator. It inhibits the growth of fibroblasts expressing the p53R175 mutation with an IC50 of 8 nM, showing no inhibition for p53 wild-type cells. This compound has demonstrated significant inhibition of tumor growth in TOV112D (p53R175H) xenografts in vivo, providing evidence for allele-specific p53 mutant reactivation.
  • Mutant p53R175 reactivator with IC50 of 8 nM against p53R175 mutation
  • No inhibition for p53 wild-type cells
  • High sensitivity for MEF-p53R172H/R172H cell line
  • Inhibits tumor growth in TOV112D (p53R175H) xenografts in vivo
  • Provides in vivo evidence for allele-specific p53 mutant reactivation
  • CC50 < 0.02 μM for Caco-2 cell toxicity
  • IC50 < 0.02 μM for Caco-2 cell SARS-CoV-2 cytotoxicity inhibition
  • EC50 0.005 μM for TOV112D cell growth inhibition
Catalog No. 50-003-87865 Supplier Medchemexpress LLC Supplier No. HY18634100MG
NSC319726 (ZMC1) is a mutant p53R175 reactivator. It inhibits the growth of fibroblasts expressing the p53R175 mutation with an IC50 of 8 nM, showing no inhibition for p53 wild-type cells. This compound has demonstrated significant inhibition of tumor growth in TOV112D (p53R175H) xenografts in vivo, providing evidence for allele-specific p53 mutant reactivation.
  • Mutant p53R175 reactivator with IC50 of 8 nM against p53R175 mutation
  • No inhibition for p53 wild-type cells
  • High sensitivity for MEF-p53R172H/R172H cell line
  • Inhibits tumor growth in TOV112D (p53R175H) xenografts in vivo
  • Provides in vivo evidence for allele-specific p53 mutant reactivation
  • CC50 < 0.02 μM for Caco-2 cell toxicity
  • IC50 < 0.02 μM for Caco-2 cell SARS-CoV-2 cytotoxicity inhibition
  • EC50 0.005 μM for TOV112D cell growth inhibition