Learn More
Medchemexpress LLC HY-19726 5mg Medchemexpress, NSC59984 CAS:803647-40-7 Purity:>98%
SDP
Catalog No. 502082169
Encompass_Preferred
Medchemexpress, HY-19726 5mg NSC59984 CAS:803647-40-7 NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2169 Supplier Medchemexpress LLC Supplier No. HY197265MG
Medchemexpress, HY-19726 5mg NSC59984 CAS:803647-40-7 NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.