Learn More
TARGETMOL CHEMICALS INC NSC632839 25MG
SDP
Catalog No. 502580051
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NSC-632839 (Ubiquitin Isopeptidase Inhibitor II) is a nonselective isopeptidase inhibitor which inhibits USP2 (EC50 45±4 uM) USP7 (EC50 37±1 uM) and SENP2 (EC50 9.8±1.8 uM). purity: 99%
Catalog No. 50-258-0051 Supplier TARGETMOL CHEMICALS INC Supplier No. T395125MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NSC-632839 (Ubiquitin Isopeptidase Inhibitor II) is a nonselective isopeptidase inhibitor which inhibits USP2 (EC50 45±4 uM) USP7 (EC50 37±1 uM) and SENP2 (EC50 9.8±1.8 uM). purity: 99%