Learn More
Medchemexpress LLC HY-100708 5mg Medchemexpress, NSC632839 CAS:157654-67-6 Purity:>98%
SDP
Catalog No. 501931922
Encompass_Preferred
Medchemexpress, HY-100708 5mg NSC632839 CAS:157654-67-6 NSC632839 is a nonselective isopeptidase inhibitor, which inhibits USP2 , USP7 , and SENP2 with EC 50 s of 45±4 μM, 37±1 μM, and 9.8±1.8 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1922 Supplier Medchemexpress LLC Supplier No. HY1007085MG
Add to Cart
Add to Cart
Medchemexpress, HY-100708 5mg NSC632839 CAS:157654-67-6 NSC632839 is a nonselective isopeptidase inhibitor, which inhibits USP2 , USP7 , and SENP2 with EC 50 s of 45±4 μM, 37±1 μM, and 9.8±1.8 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.