Learn More
Medchemexpress LLC Nsc639828 | 134742-26-0 | 99.8% | C18H13BrClN5O3 | 100 MG
SDP
Catalog No. 5000355264
Encompass_Preferred
Nsc639828 is a potent inhibitor of DNA polymerase α with an IC50 of 70 μM. It exhibits high antitumor activity and has potential for researching cancer disease.
  • Potent inhibitor of DNA polymerase α
  • Ic50 of 70 μM
  • Exhibits high antitumor activity
  • Potential for researching cancer disease
Catalog No. 50-003-55264 Supplier Medchemexpress LLC Supplier No. HY145330100MG
Nsc639828 is a potent inhibitor of DNA polymerase α with an IC50 of 70 μM. It exhibits high antitumor activity and has potential for researching cancer disease.
  • Potent inhibitor of DNA polymerase α
  • Ic50 of 70 μM
  • Exhibits high antitumor activity
  • Potential for researching cancer disease