Learn More
Medchemexpress LLC NSC668394 100mg | 382605-72-3 | 452.1 g/mol | C17H12Br2N2O3 | 100 MG
SDP
Catalog No. 5000229277
Encompass_Preferred
NSC668394 is a small-molecule inhibitor of ezrin Thr567 phosphorylation offered for research use. It is supplied as an orange to red solid with molecular formula C17H12Br2N2O3, molecular weight 452.1 g/mol, and reported purity of 99.31%. The compound is available in multiple pack sizes, including 100 MG, and has recommended storage conditions for powder and solutions to preserve stability.
  • Inhibits ezrin Thr567 phosphorylation for cellular and biochemical studies.
  • High reported purity (99.31%) suitable for research applications.
  • Orange to red solid physical form for easy handling.
  • Molecular weight 452.1 g/mol; formula C17H12Br2N2O3.
  • Available in multiple pack sizes, including 100 MG.
  • Powder storage: -20°C for long-term stability; in solvent: -80°C recommended for extended storage.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-29277 Supplier Medchemexpress LLC Supplier No. HY115492100MG
NSC668394 is a small-molecule inhibitor of ezrin Thr567 phosphorylation offered for research use. It is supplied as an orange to red solid with molecular formula C17H12Br2N2O3, molecular weight 452.1 g/mol, and reported purity of 99.31%. The compound is available in multiple pack sizes, including 100 MG, and has recommended storage conditions for powder and solutions to preserve stability.
  • Inhibits ezrin Thr567 phosphorylation for cellular and biochemical studies.
  • High reported purity (99.31%) suitable for research applications.
  • Orange to red solid physical form for easy handling.
  • Molecular weight 452.1 g/mol; formula C17H12Br2N2O3.
  • Available in multiple pack sizes, including 100 MG.
  • Powder storage: -20°C for long-term stability; in solvent: -80°C recommended for extended storage.
  • For research use only; not for human or veterinary use.