Learn More
Medchemexpress LLC NSC668394 10mg | 382605-72-3 | 452.10 g·mol^-1 | C17H12Br2N2O3 | 10 MG
SDP
Catalog No. 5000229313
Encompass_Preferred
NSC668394 is a small-molecule ezrin phosphorylation inhibitor used in preclinical research to study ezrin-dependent signaling and metastatic processes. It binds ezrin and reduces Thr567 phosphorylation, affecting cytoskeleton-associated signaling pathways.
  • Inhibits ezrin Thr567 phosphorylation and ezrin-mediated signaling.
  • Suitable for in vitro, cell-based studies due to cell permeability.
  • High reported purity (99.31%) for reliable experimental results.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 452.10 g·mol⁻¹; formula C17H12Br2N2O3.
Catalog No. 50-002-29313 Supplier Medchemexpress LLC Supplier No. HY11549210MG
Add to Cart
Add to Cart
NSC668394 is a small-molecule ezrin phosphorylation inhibitor used in preclinical research to study ezrin-dependent signaling and metastatic processes. It binds ezrin and reduces Thr567 phosphorylation, affecting cytoskeleton-associated signaling pathways.
  • Inhibits ezrin Thr567 phosphorylation and ezrin-mediated signaling.
  • Suitable for in vitro, cell-based studies due to cell permeability.
  • High reported purity (99.31%) for reliable experimental results.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 452.10 g·mol⁻¹; formula C17H12Br2N2O3.