Learn More
Selleck Chemical LLC NSC697923-10mg
SDP
Catalog No. 502389748
Encompass_Preferred
NSC697923 is a cell-permeable and selective inhibitor of the Ub-conjugating enzyme (E2) complex Ubc13-Uev1A.
Catalog No. 50-238-9748 Supplier Selleck Chemical LLC Supplier No. S714210MG
NSC697923 is a cell-permeable and selective inhibitor of the Ub-conjugating enzyme (E2) complex Ubc13-Uev1A.