Learn More
AdipoGen NSC697923 (25 mg)
SDP
Supplier:  AdipoGen AGCR13519M025
Encompass_Preferred
Chemical. CAS: 343351-67-7. Formula: C11H9NO5S. MW: 267.3. Selective Ub-conjugating enzyme (E2) complex Ubc13-Uev1A inhibitor. Inhibits the formation of the Ubc13~Ub conjugate. NF-kappaB activation inhibitor. Inhibits proliferation and survival of ABC-DLBCL and GCB-DLBCL (diffuse large B cell lymphoma) cells.
Catalog No. 50-205-3902