Learn More
Enzo Life Sciences NSC697923 (5mg). CAS: 343351-67-7
SDP
Catalog No. 502013445
Encompass_Preferred
Alternative name: 2-[(4-Methylphenyl)sulfonyl]-5-nitro-furan, 2-Nitro-5-tosylfuran . Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3445 Supplier Enzo Life Sciences Supplier No. ENZCHM1430005
Alternative name: 2-[(4-Methylphenyl)sulfonyl]-5-nitro-furan, 2-Nitro-5-tosylfuran . Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.