Learn More
Medchemexpress LLC HY-100470 5mg Medchemexpress, NSC781406 CAS:1676893-24-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004705MG
Encompass_Preferred
Medchemexpress, HY-100470 5mg NSC781406 CAS:1676893-24-5 NSC781406 is a highly potent PI3K and mTOR inhibitor with an IC 50 of 2 nM for PI3Kα. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1846