Learn More
Eurofins Discoverx NSD1, 50 µg. (EPI041)
Supplier:  Eurofins Discoverx EPI041
Encompass
NSD1 is purified, untagged recombinant NSD1 enzyme. This enzyme contains amino acids 79-447. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5259