Learn More
ABclonal Technology NSDHL Rabbit mAb
SDP
Catalog No. 5000008091
Encompass_Preferred
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis Mutations in this gene are associated with CHILD syndrome which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis and typically lethal in males Alternatively spliced transcript variants with differing 5 UTR have been found for this gene
Catalog No. 50-000-08091 Supplier ABclonal Technology Supplier No. A21152100UL
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis Mutations in this gene are associated with CHILD syndrome which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis and typically lethal in males Alternatively spliced transcript variants with differing 5 UTR have been found for this gene