Learn More
Genetex Inc NSE Monoclonal Antibody [MSVA-451M] HistoMAX™ (GTX04388) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04388
Encompass_Preferred
The NSE antibody [MSVA-451M] HistoMAX™ (GTX04388) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human adrenal pheochromocytoma tissue using GTX04388 NSE antibody [MSVA-451M] HistoMAX. Strong NSE immunostaining in an adrenal pheochromocytoma.
Catalog No. NC3666326