Learn More
Enzo Life Sciences NSF monoclonal antibody (9G7-3) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMSV020D
Encompass_Preferred
Clone: 9G7-3. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant bovine NSF. UniProt ID: P46459 (human). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Dog, Chicken, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7698