Learn More
SPEX CERTIPREP LLC 3P Minerals QC CRM, 4 x 500 mL
Catalog No. NC0070254
Encompass_Preferred
(NSI by Zeptometrix) A 500 mL ready-to-use sample packaged in a HDPE bottle to be analyzed for the following: Potassium 4.0 - 40 mg/L, Sodium 10 - 100 mg/L, Chloride 35 - 275 mg/L, Sulfate 5.0 - 125 mg/L, Fluoride 0.4 - 4 mg/L, TDS at 180C 140 - 800 mg/L, Conductivity 200 - 1200 umhos/cm, Alkalinity 25 - 400 mg/L.
Catalog No. NC0070254 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI136C
(NSI by Zeptometrix) A 500 mL ready-to-use sample packaged in a HDPE bottle to be analyzed for the following: Potassium 4.0 - 40 mg/L, Sodium 10 - 100 mg/L, Chloride 35 - 275 mg/L, Sulfate 5.0 - 125 mg/L, Fluoride 0.4 - 4 mg/L, TDS at 180C 140 - 800 mg/L, Conductivity 200 - 1200 umhos/cm, Alkalinity 25 - 400 mg/L.