Learn More
New England Biolabs, Inc. NsiI – 1000 units
SDP
Catalog No. 50811975
Encompass
A restriction endonuclease that recognizes the sequence A_TGCA^T.
Catalog No. 50-811-975 Supplier New England Biolabs, Inc. Supplier No. R0127S
A restriction endonuclease that recognizes the sequence A_TGCA^T.