Learn More
New England Biolabs, Inc. NsiI – 5000 units
SDP
Catalog No. 50811974
Encompass
A restriction endonuclease that recognizes the sequence A_TGCA^T.
Catalog No. 50-811-974 Supplier New England Biolabs, Inc. Supplier No. R0127L
A restriction endonuclease that recognizes the sequence A_TGCA^T.