Learn More
Cayman Chemical nsIrItIdeacetate 10mg
SDP
Catalog No. 502949886
Encompass_Preferred
A natriuretic peptide; increases ex vivo A23187-induced vasorelaxation in porcine hearts isolated from a model of acute coronary occlusion at 2 UG/kg bolus dose followed by 0.01 UG/kg per minute infusion
Catalog No. 50-294-9886 Supplier Cayman Chemical Supplier No. 3453210 MG
A natriuretic peptide; increases ex vivo A23187-induced vasorelaxation in porcine hearts isolated from a model of acute coronary occlusion at 2 UG/kg bolus dose followed by 0.01 UG/kg per minute infusion