Learn More
Aviva Systems Biology Corp NSL1 Antibody - C-terminal region (ARP62184_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62184_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NSL1. It was validated on Western Blot. . Alternate names: C1orf48, DC8, DKFZp566O1646, MIS14ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-287-603