Learn More
Aviva Systems Biology Corp NSMCE2 Antibody - middle region (ARP55690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NSMCE2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NSMCE2. Alternate names: C8orf36, FLJ32440, MMS21, NSE2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-61580