Learn More
Medchemexpress LLC Nsp-dmae-nhs | 194357-64-7 | 99.5% | 1 MG
SDP
Catalog No. 5000410414
Encompass_Preferred
NSP-DMAE-NHS is an acridinium ester chemiluminescence reagent containing an N-sulfopropyl group. It is useful in the clinical diagnostics industry, especially in automated immunochemistry analyzers. The product is for research use only and not sold to patients.
  • Acridinium ester chemiluminescence reagent
  • Contains n-sulfopropyl group
  • Useful in the clinical diagnostics industry
  • Applicable in automated immunochemistry analyzers
Catalog No. 50-004-10414 Supplier Medchemexpress LLC Supplier No. HYD08921MG
NSP-DMAE-NHS is an acridinium ester chemiluminescence reagent containing an N-sulfopropyl group. It is useful in the clinical diagnostics industry, especially in automated immunochemistry analyzers. The product is for research use only and not sold to patients.
  • Acridinium ester chemiluminescence reagent
  • Contains n-sulfopropyl group
  • Useful in the clinical diagnostics industry
  • Applicable in automated immunochemistry analyzers