Learn More
Medchemexpress LLC NSP-SA-NHS | 199293-83-9 | 99.65% | 100 MG
SDP
Catalog No. 5000409875
Encompass_Preferred
NSP-SA-NHS is an acridinium ester used for chemiluminescent immunoassay. A rapid and sensitive chemiluminescent immunoassay for Bisphenol A (BPA) has been developed using NSP-SA-NHS-labeled. It is for research use only.
  • Used for chemiluminescent immunoassay.
  • Enabled the development of a rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA).
Catalog No. 50-004-09875 Supplier Medchemexpress LLC Supplier No. HYD0893100MG
NSP-SA-NHS is an acridinium ester used for chemiluminescent immunoassay. A rapid and sensitive chemiluminescent immunoassay for Bisphenol A (BPA) has been developed using NSP-SA-NHS-labeled. It is for research use only.
  • Used for chemiluminescent immunoassay.
  • Enabled the development of a rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA).