Learn More
Medchemexpress LLC NSP-SA-NHS | 199293-83-9 | 99.65% | 50 MG
SDP
Catalog No. 5000410458
Encompass_Preferred
NSP-SA-NHS is an acridinium ester that can be used for chemiluminescent immunoassay. A rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA) with NSP-SA-NHS-labeled has been developed.
  • Application: Chemiluminescent immunoassay.
  • Specific use: Developed for a rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA).
Catalog No. 50-004-10458 Supplier Medchemexpress LLC Supplier No. HYD089350MG
NSP-SA-NHS is an acridinium ester that can be used for chemiluminescent immunoassay. A rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA) with NSP-SA-NHS-labeled has been developed.
  • Application: Chemiluminescent immunoassay.
  • Specific use: Developed for a rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA).