Learn More
Medchemexpress LLC NSP-SA-NHS | 199293-83-9 | 99.7% | 1 MG
SDP
Catalog No. 5000409872
Encompass_Preferred
NSP-SA-NHS is an acridinium ester used for chemiluminescent immunoassay. It enables rapid and sensitive detection, making it suitable for developing immunoassays such as for Bisphenol A (BPA). This product is for research use only.
  • Solid appearance
  • Light yellow to yellow color
  • Ships at room temperature
  • Store at -20°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
Catalog No. 50-004-09872 Supplier Medchemexpress LLC Supplier No. HYD08931MG
NSP-SA-NHS is an acridinium ester used for chemiluminescent immunoassay. It enables rapid and sensitive detection, making it suitable for developing immunoassays such as for Bisphenol A (BPA). This product is for research use only.
  • Solid appearance
  • Light yellow to yellow color
  • Ships at room temperature
  • Store at -20°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light