Learn More
Medchemexpress LLC NSP-SA-NHS | 199293-83-9 | 99.7% | 25 MG
SDP
Catalog No. 5000413563
Encompass_Preferred
NSP-SA-NHS is an acridinium ester designed for chemiluminescent immunoassay. It enables the development of rapid and sensitive immunoassays, such as for Bisphenol A (BPA) detection.
  • Can be used for chemiluminescent immunoassay.
  • A rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA) with NSP-SA-NHS-labeled has been developed.
Catalog No. 50-004-13563 Supplier Medchemexpress LLC Supplier No. 25MGHYD0893
NSP-SA-NHS is an acridinium ester designed for chemiluminescent immunoassay. It enables the development of rapid and sensitive immunoassays, such as for Bisphenol A (BPA) detection.
  • Can be used for chemiluminescent immunoassay.
  • A rapid and sensitive chemiluminescent immunoassay of Bisphenol A (BPA) with NSP-SA-NHS-labeled has been developed.