Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nsp-sa-nhs | 199293-83-9 | 99.7% | 681.73 | C32H31N3O10S2 | 10 MG
SDP

Catalog No. 5000297670
Encompass_Preferred

Acridinium ester reagent for chemiluminescent labeling in immunoassays. It is provided as a high-purity solid for research use and is intended to enable sensitive detection when conjugated to analytes in chemiluminescent assay workflows.

  • Used for chemiluminescent immunoassay labeling.
  • High purity (≈99.7%) for sensitive detection.
  • Molecular weight 681.73 g·mol-1 and formula C32H31N3O10S2.
  • CAS number 199293-83-9 for unambiguous identification.
  • Supplied in milligram quantities suitable for conjugation workflows.
  • Store sealed, protected from moisture and light, preferably at -20°C.

Catalog No. 50-002-97670 Supplier Medchemexpress LLC Supplier No. HYD089310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acridinium ester reagent for chemiluminescent labeling in immunoassays. It is provided as a high-purity solid for research use and is intended to enable sensitive detection when conjugated to analytes in chemiluminescent assay workflows.

  • Used for chemiluminescent immunoassay labeling.
  • High purity (≈99.7%) for sensitive detection.
  • Molecular weight 681.73 g·mol-1 and formula C32H31N3O10S2.
  • CAS number 199293-83-9 for unambiguous identification.
  • Supplied in milligram quantities suitable for conjugation workflows.
  • Store sealed, protected from moisture and light, preferably at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.