Learn More
TARGETMOL CHEMICALS INC NSP-SA-NHS 5MG
SDP
Catalog No. 502878579
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. NSP-SA-NHS is a chemiluminescent acridinium ester that can be used for immunoassays. Purity 98.42%
Catalog No. 50-287-8579 Supplier TARGETMOL CHEMICALS INC Supplier No. T190405MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. NSP-SA-NHS is a chemiluminescent acridinium ester that can be used for immunoassays. Purity 98.42%