Learn More
Cayman Chemical N-Succinimidyl-S-acetylthioacetate, Purity: ≥90%, CAS: 76931-93-6, 50 mg
SDP
Catalog No. 502914881
Encompass_Preferred
A protein-modifying agent; introduces sulfhydryl groups to proteins; used in conjunction with the protein cross-linking reagent SMCC in the conjugation of proteins with IgG antibodies for use in ELISAs
Catalog No. 50-291-4881 Supplier Cayman Chemical Supplier No. 3692850 MG
A protein-modifying agent; introduces sulfhydryl groups to proteins; used in conjunction with the protein cross-linking reagent SMCC in the conjugation of proteins with IgG antibodies for use in ELISAs