Learn More
Aviva Systems Biology Corp NSUN2 Antibody - C-terminal region (ARP48811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NSUN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NSUN2. Alternate names: FLJ20303, MISU, SAKI, TRM4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53562