Learn More
Aviva Systems Biology Corp NSUN4 Antibody - N-terminal region (ARP49196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NSUN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NSUN4. Alternate names: MGC22960, RP4-603I14.2, SHTAPApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60039