Learn More
Aviva Systems Biology Corp NSUN5C Antibody - middle region (ARP48960_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48960_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NSUN5C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NSUN5C. Alternate names: DKFZp434K058, DKFZp666P104, FLJ11626, MGC129801, MGC15057, NOL1R2, WBSCR20B, WBSCR20C, NSUN5CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59977