Learn More
Medchemexpress LLC HY-P7272 10ug Medchemexpress, NT-4, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP727210UG
Encompass_Preferred
Medchemexpress, HY-P7272 10ug NT-4, Human CAS: Recombinant Human Neurotrophin-4 is a member of neurotrophins family, binding with two distinct receptors: TrkB, high affinity receptor and p75 low-affinity neurotrophin receptor (p75NTR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0590