Learn More
Medchemexpress LLC NT-4 Mouse 10ug
SDP
Catalog No. 5000182381
Encompass_Preferred
NT-4 Protein Mouse is a member of neurotrophins family binding with two distinct receptors TrkB high affinity receptor and p75 low-affinity neurotrophin receptor (p75NTR)
Catalog No. 50-001-82381 Supplier Medchemexpress LLC Supplier No. HYP727310UG
NT-4 Protein Mouse is a member of neurotrophins family binding with two distinct receptors TrkB high affinity receptor and p75 low-affinity neurotrophin receptor (p75NTR)