Learn More
New England Biolabs, Inc. Nt.AlwI – 500 units
SDP
Catalog No. 50812370
Encompass
A restriction endonuclease that recognizes the sequence GGATCNNNN^N. Nt.AlwI is a nicking endonuclease that cleaves only one strand of DNA on a double-stranded DNA substrate. It is an engineered derivative of AlwI which catalyzes a single strand break 4 bases beyond the 3' end of the recognition sequence on the top strand.
Catalog No. 50-812-370 Supplier New England Biolabs, Inc. Supplier No. R0627S
A restriction endonuclease that recognizes the sequence GGATCNNNN^N. Nt.AlwI is a nicking endonuclease that cleaves only one strand of DNA on a double-stranded DNA substrate. It is an engineered derivative of AlwI which catalyzes a single strand break 4 bases beyond the 3' end of the recognition sequence on the top strand.