Learn More
New England Biolabs, Inc. Nt.BbvCI – 1000 units
SDP
Catalog No. 50812378
Encompass
A restriction endonuclease that recognizes the sequence CC^TCAGC.
Catalog No. 50-812-378 Supplier New England Biolabs, Inc. Supplier No. R0632S
A restriction endonuclease that recognizes the sequence CC^TCAGC.