Learn More
New England Biolabs, Inc. Nt.BbvCI – 5000 units
SDP
Catalog No. 50812377
Encompass
A restriction endonuclease that recognizes the sequence CC^TCAGC.
Catalog No. 50-812-377 Supplier New England Biolabs, Inc. Supplier No. R0632L
A restriction endonuclease that recognizes the sequence CC^TCAGC.