Learn More
New England Biolabs, Inc. Nt.BspQI – 1000 units
SDP
Catalog No. 50995264
Encompass
A restriction endonuclease that recognizes the sequence GCTCTTCN^.
Catalog No. 50-995-264 Supplier New England Biolabs, Inc. Supplier No. R0644S
A restriction endonuclease that recognizes the sequence GCTCTTCN^.