Learn More
New England Biolabs, Inc. Nt.BstNBI – 1000 units
SDP
Catalog No. 50812336
Encompass
A restriction endonuclease that recognizes the sequence GAGTCNNNN^N.
Catalog No. 50-812-336 Supplier New England Biolabs, Inc. Supplier No. R0607S
A restriction endonuclease that recognizes the sequence GAGTCNNNN^N.