Learn More
New England Biolabs, Inc. Nt.BstNBI – 5000 units
SDP
Catalog No. 50812335
Encompass
A restriction endonuclease that recognizes the sequence GAGTCNNNN^N.
Catalog No. 50-812-335 Supplier New England Biolabs, Inc. Supplier No. R0607L
A restriction endonuclease that recognizes the sequence GAGTCNNNN^N.