Learn More
New England Biolabs, Inc. Nt.CviPII – 40 units
SDP
Catalog No. 502108566
Encompass
A restriction endonuclease that recognizes the sequence ^CCD. Nt.CviPII is a nicking endonuclease that cleaves only one strand of DNA on a double-stranded DNA substrate. The final product on pUC19 is an array of bands from 25 to 200 bp. CCT is cut less efficiently than CCG and CCA. Some of the CCT sites are not cleaved. (Based on NEB observations.)
Catalog No. 50-210-8566 Supplier New England Biolabs, Inc. Supplier No. R0626S
A restriction endonuclease that recognizes the sequence ^CCD. Nt.CviPII is a nicking endonuclease that cleaves only one strand of DNA on a double-stranded DNA substrate. The final product on pUC19 is an array of bands from 25 to 200 bp. CCT is cut less efficiently than CCG and CCA. Some of the CCT sites are not cleaved. (Based on NEB observations.)