Learn More
Genscript Corporation NT-proBNP 3B42HC
Catalog No. 5000057715
Encompass
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction They can be used for diagnosis of congestive heart failure (CHF)
Catalog No. 50-000-57715 Supplier Genscript Corporation Supplier No. V007041
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction They can be used for diagnosis of congestive heart failure (CHF)