Learn More
Genscript Corporation NT-proBNP 3B42HC
Catalog No. 5000058408
Encompass
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction They can be used for diagnosis of congestive heart failure (CHF)
Catalog No. 50-000-58408 Supplier Genscript Corporation Supplier No. V00704100
Add to Cart
Add to Cart
Request a Quote
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction They can be used for diagnosis of congestive heart failure (CHF)