Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NT-proBNP (Human) ELISA Kit

Catalog No. 89100802 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum
CCS, Plasma, Serum, Tissue, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-802 CCS, Plasma, Serum
89-100-381 CCS, Plasma, Serum, Tissue, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-802 Supplier Abnova™ Supplier No. KA3099
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-human NT-proBNP antibody, 96 (8x12) wells
  • Human NT-proBNP standards, 10ng/tube x 2
  • Sample/Standard diluent buffer, 30mL
  • HRP conjugated anti-human NT-proBNP antibody (Concentrated): Dilution: 1:100 (store at 4°C, do not store at -20°C),150μL
  • Antibody diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer: (25x): Dilution: 1:25, 30mL

Use to measure quantitatively human NT-proBNP.

The kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate HRP
Cross Reactivity Human
Description NT-proBNP (Human) ELISA Kit is intended for the quantitative measurement of human NT-proBNP.
Detection Limit 2pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-human NT-proBNP antibody (96 (8 x 12) wells, Human NT-proBNP standards: (1μg/mL): 20μL x 2, Sample/Standard diluent buffer (30mL), HRP conjugated anti-human NT-proBNP antibody (Concentrated): Dilution: 1:100 (150μL), Antibody diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer: (25x), Dilution: 1:25 (30mL)
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 100 μL
Sensitivity 2 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type NT-proBNP (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.